Läs mer
5,6-Dihydrouridine, TRC
Produktkod. 30134025
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134025 50 mg
30006113 5 mg
30005540 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134025 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44966850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5627-05-4
Molekylformel C9H14N2O6
Molekylvikt (g/mol) 246.22
Synonym Dihydro-1-β-D-ribofuranosyl-2,4(1H,3H)-pyrimidinedione, Dihydrouridine, 1-β-D-Ribofuranosyl-hydrouracil;
LEDER O=C(NC1=O)CCN1[C@H]2[C@H](O)[C@H](O)[C@@H](CO)O2
Specifikationer
Kemiskt namn eller material 5,6-Dihydrouridine
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C9H14N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h4,6-8,12,14-15H,1-3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1
Formel vikt 246.0852
Procent renhet >95%