Läs mer
5,6-Dihydroxyindole-2-carboxylic Acid, TRC
Produktkod. 30005922
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005922 10 mg
30005036 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45282510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4790-08-3
Molekylformel C9 H7 N O4
Molekylvikt (g/mol) 193.16
Synonym 2-Carboxy-5,6-dihydroxyindole, 5,6-Dihydroxy-1H-indole-2-carboxylic Acid, 5,6-Dihydroxy-2-carboxyindole
IUPAC-namn 5,6-dihydroxy-1H-indole-2-carboxylic acid
LEDER OC(=O)c1cc2cc(O)c(O)cc2[nH]1
Specifikationer
Kemiskt namn eller material 5,6-Dihydroxyindole-2-carboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C9H7NO4/c11-7-2-4-1-6(9(13)14)10-5(4)3-8(7)12/h1-3,10-12H,(H,13,14)
Formel vikt 193.0375
Procent renhet >95%