missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5,6-Dimethoxy-2-phenylindole, 97%
Click to view available options
Quantity:
0.1 g
0.5 g
Specifications
Quantity | 0.1 g |
Formula Weight | 253.3 |
Percent Purity | 97% |
Chemical Name or Material | 5,6-Dimethoxy-2-phenylindole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'