Läs mer
5,6-Dimethylbenzotriazole Hydrate, TRC
Produktkod. 17972165
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17972165 1 g
17901725 5 g
17920595 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17972165 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4622751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1354973-50-4
Molekylformel C8 H9 N3 . H2 O
Molekylvikt (g/mol) 165.19
Synonym 5,6-Dimethyl-1H-benzotriazole monohydrate, 5,6-Dimethyl-1,2,3-benzotriazole Hydrate, 5,6-Dimethylbenzotriazole Hydrate, NSC 62005 Hydrate
IUPAC-namn 5,6-dimethyl-1H-benzotriazole;hydrate
LEDER O.Cc1cc2nn[nH]c2cc1C
Specifikationer
Kemiskt namn eller material 5,6-Dimethyl-1H-benzotriazole monohydrate
Kvantitet 1 g
InChI formel InChI=1S/C8H9N3.H2O/c1-5-3-7-8(4-6(5)2)10-11-9-7;/h3-4H,1-2H3,(H,9,10,11);1H2
Formel vikt 165.0902