Läs mer
5,7-Dichloro-8-Quinolinol Glucuronide, TRC
Produktkod. 30240959
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30240959 25 mg -
30240921 2.5 mg 2.5mg
30240799 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240959 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43630025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40951-47-1
Molekylformel C15 H13 Cl2 N O7
Molekylvikt (g/mol) 390.172
Synonym 5,7-Dichloro-8-Quinolinyl β-D-Glucopyranosiduronic Acid;
IUPAC-namn (2S,3S,4S,5R,6S)-6-(5,7-dichloroquinolin-8-yl)oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER O[C@H]1[C@H](Oc2c(Cl)cc(Cl)c3cccnc23)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O
Specifikationer
Kemiskt namn eller material 5,7-Dichloro-8-Quinolinol Glucuronide
Kvantitet 25 mg
InChI formel InChI=1S/C15H13Cl2NO7/c16-6-4-7(17)12(8-5(6)2-1-3-18-8)24-15-11(21)9(19)10(20)13(25-15)14(22)23/h1-4,9-11,13,15,19-21H,(H,22,23)/t9-,10-,11+,13-,15+/m0/s1
Formel vikt 389.007