missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloropyrazolo[1,5-a]pyrimidine, 95%, ACROS Organics™

Brand:  Acros Organics 454132500

Additional Details : CAS Number : 57489-77-7 Weight : 0.00025kg


Quantity 250mg

 View more versions of this product

Product Code. 15214328

  • 1457.00 SEK / 250mg

Estimated Shipment
30-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

57489-77-7
188.02
5,7-dichloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 5,7-dichloro, 5,7-dichloro-pyrazolo 1,5-a pyrimidine, pubchem14638, 5,7-dichloropyrazolo 1,5-a pyri-midine, 5, 7-dichloro-pyrazolo 1,5-a pyrimidine, 5,7-bis chloranyl pyrazolo 1,5-a pyrimidine
5,7-dichloropyrazolo[1,5-a]pyrimidine
C6H3Cl2N3
JMTFWCYVZOFHLR-UHFFFAOYSA-N
11074154
C1=C2N=C(C=C(N2N=C1)Cl)Cl

Specifications

5,7-Dichloropyrazolo[1,5-a]pyrimidine
75.0°C to 76.5°C
57489-77-7
C6H3Cl2N3
JMTFWCYVZOFHLR-UHFFFAOYSA-N
5,7-dichloropyrazolo[1,5-a]pyrimidine
11074154
95%
Authentic
250mg
94% min. (GC)
5,7-dichloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 5,7-dichloro, 5,7-dichloro-pyrazolo 1,5-a pyrimidine, pubchem14638, 5,7-dichloropyrazolo 1,5-a pyri-midine, 5, 7-dichloro-pyrazolo 1,5-a pyrimidine, 5,7-bis chloranyl pyrazolo 1,5-a pyrimidine
C1=C2N=C(C=C(N2N=C1)Cl)Cl
188.02
188.02
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5,7-Dichloropyrazolo¢1,5-a!pyrimidine, 95%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents