Läs mer
5,8-Epoxy-13-cis Retinoic Acid (Mixture of Diastereomers), TRC
Produktkod. 30369798
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30369798 5 mg
1 options
Produktkod. 30369798 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5898205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5,8-Epoxy-13-cis Retinoic Acid (Mixture of Diastereomers)
CAS 112018-12-9
Molekylformel C20H28O3
Kvantitet 5 mg
Synonym (2Z,4E,6E)-7-(2,4,5,6,7,7a-Hexahydro-4,4,7a-trimethyl-2-benzofuranyl)-3-methyl-2,4,6-octatrienoic Acid, 13-cis-5,8-Epoxy-5,8-dihydroretinoic Acid
InChI formel InChI=1S/C20H28O3/c1-14(12-18(21)22)8-6-9-15(2)16-13-17-19(3,4)10-7-11-20(17,5)23-16/h6,8-9,12-13,16H,7,10-11H2,1-5H3,(H,21,22)/b8-6+,14-12-,15-9+
LEDER C/C(=C/C(=O)O)/C=C/C=C(\C)/C1C=C2C(CCCC2(O1)C)(C)C
Molekylvikt (g/mol) 316.43