Läs mer
5-Acetamino-4-hydroxy-2-(4-nitro-phenoxy)-6-(1,2,3-trihydroxy-propyl)-tetrahydro-pyran-2-carboxylic Acid Methyl Ester, TRC
Produktkod. 30315539
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30315539 10 mg
1 options
Produktkod. 30315539 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA15844510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Acetamino-4-hydroxy-2-(4-nitro-phenoxy)-6-(1,2,3-trihydroxy-propyl)-tetrahydro-pyran-2-carboxylic Acid Methyl Ester
CAS 59694-35-8
Molekylformel C18H24N2O11
Kvantitet 10 mg
Synonym N-Acetyl-2-O-(4-nitrophenyl)-alpha-neuraminic Acid Methyl Ester
InChI formel InChI=1S/C18H24N2O11/c1-9(22)19-14-12(23)7-18(17(26)29-2,31-16(14)15(25)13(24)8-21)30-11-5-3-10(4-6-11)20(27)28/h3-6,12-16,21,23-25H,7-8H2,1-2H3,(H,19,22)
LEDER COC(=O)C1(CC(O)C(NC(=O)C)C(O1)C(O)C(O)CO)Oc2ccc(cc2)[N+](=O)[O-]
IUPAC-namn methyl 5-acetamido-4-hydroxy-2-(4-nitrophenoxy)-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylate
Molekylvikt (g/mol) 444.39
Formel vikt 444.138