missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-amino-4-methylthiazole, 97+%, Thermo Scientific Chemicals

Catalog Number p-7045011
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
30748-47-1
C6H8N2OS
156.203
MFCD00051952
PKUKCASRNJIQNU-UHFFFAOYSA-N
2-amino-4-methyl-5-acetylthiazole, 1-2-amino-4-methylthiazol-5-yl ethanone, 1-2-amino-4-methyl-1,3-thiazol-5-yl ethanone, 5-acetyl-2-amino-4-methylthiazole, 1-2-amino-4-methyl-1,3-thiazol-5-yl ethan-1-one, 1-2-amino-4-methyl-thiazol-5-yl-ethanone, 5-acetyl-2-amino-4-methyl-1,3-thiazole, ethanone, 1-2-amino-4-methyl-5-thiazolyl, 1-2-amino-4-methyl-1,3-thiazol-5-yl-1-ethanone, 5-acetyl-4-methyl-2-thiazolamine
720882
1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethanone
CC1=C(SC(=N1)N)C(=O)C
This item is not returnable. View return policy
30748-47-1
C6H8N2OS
156.203
MFCD00051952
PKUKCASRNJIQNU-UHFFFAOYSA-N
2-amino-4-methyl-5-acetylthiazole, 1-2-amino-4-methylthiazol-5-yl ethanone, 1-2-amino-4-methyl-1,3-thiazol-5-yl ethanone, 5-acetyl-2-amino-4-methylthiazole, 1-2-amino-4-methyl-1,3-thiazol-5-yl ethan-1-one, 1-2-amino-4-methyl-thiazol-5-yl-ethanone, 5-acetyl-2-amino-4-methyl-1,3-thiazole, ethanone, 1-2-amino-4-methyl-5-thiazolyl, 1-2-amino-4-methyl-1,3-thiazol-5-yl-1-ethanone, 5-acetyl-4-methyl-2-thiazolamine
720882
1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethanone
CC1=C(SC(=N1)N)C(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽268°C (decomposition)
Quantity 5 g
Beilstein 127111
Formula Weight 156.21
Percent Purity ≥97%
Chemical Name or Material 5-Acetyl-2-amino-4-methylthiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'