Läs mer
5-[(Acetyloxy)methyl]-3a,6,7,7a-tetrahydro-2-methyl-(3aR,5R,6S,7R,7aR)-5H-Pyrano[3,2-d]thiazole-6,7-diol Diacetate (Ester), TRC
Produktkod. 30311960
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30311960 250 mg
1 options
Produktkod. 30311960 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA187125250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-[(Acetyloxy)methyl]-3a,6,7,7a-tetrahydro-2-methyl-(3aR,5R,6S,7R,7aR)-5H-Pyrano[3,2-d]thiazole-6,7-diol Diacetate (Ester)
CAS 67109-74-4
Molekylformel C14H19NO7S
Kvantitet 250 mg
Synonym 5-[(Acetyloxy)methyl]-3a,6,7,7a-tetrahydro-2-methyl-(3aR,5R,6S,7R,7aR)-5H-Pyrano[3,2-d]thiazole-6,7-diol 6,7-Diacetate
InChI formel InChI=1S/C14H19NO7S/c1-6-15-11-13(21-9(4)18)12(20-8(3)17)10(5-19-7(2)16)22-14(11)23-6/h10-14H,5H2,1-4H3/t10?,11-,12-,13-,14-/m1/s1
LEDER CC(=O)OCC1O[C@@H]2SC(=N[C@@H]2[C@@H](OC(=O)C)[C@@H]1OC(=O)C)C
IUPAC-namn [(3aR,6S,7R,7aR)-6,7-diacetyloxy-2-methyl-5,6,7,7a-tetrahydro-3aH-pyrano[3,2-d][1,3]thiazol-5-yl]methyl acetate
Molekylvikt (g/mol) 345.37
Formel vikt 345.0882