Läs mer
5-Amino-1,3,4-thiadiazole-2-sulfonamide Hydrochloride Salt, TRC
Produktkod. 30326363
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30326363 50 mg
1 options
Produktkod. 30326363 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63036050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Amino-1,3,4-thiadiazole-2-sulfonamide Hydrochloride Salt
CAS 120208-98-2
Molekylformel C2H5ClN4O2S2
Kvantitet 50 mg
Synonym 2-Amino-1,3,4-thiadazole-5-sulfonamide Hydrochloride Salt, L 5343, NSC 22979 Hydrochloride Salt, Tio-urasin Hydrochloride Salt, Acetazolamide EP Impurity D Hydrochloride
InChI formel InChI=1S/C2H4N4O2S2.ClH/c3-1-5-6-2(9-1)10(4,7)8;/h(H2,3,5)(H2,4,7,8);1H
LEDER NC1=NN=C(S(N)(=O)=O)S1.Cl
IUPAC-namn 5-amino-1,3,4-thiadiazole-2-sulfonamide;hydrochloride
Molekylvikt (g/mol) 216.67
Formel vikt 215.9542