missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-1H-tetrazole monohydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
Solubility
Soluble in water, ethanol (1.36%).
Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
Solubility
Soluble in water, ethanol (1.36%).
Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
Specifications
Melting Point | ∼203°C (decomposition) |
pH | 3.7 |
Quantity | 25 g |
UN Number | UN1325 |
Beilstein | 108872 |
Solubility Information | Soluble in water,ethanol (1.36%). |
Formula Weight | 103.08 (85.06 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | 5-Amino-1H-tetrazole monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'