Learn More
5-Amino-2-(4-morpholinyl)pyridine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.
Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.
Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
Specifications
Melting Point | 127°C to 130°C |
Quantity | 250 mg |
Formula Weight | 179.22 |
Percent Purity | 97% |
Chemical Name or Material | 5-Amino-2-(4-morpholinyl)pyridine |