Learn More
5-Amino-2-bromobenzoic Acid, TRC
Product Code. 30309298
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
30309298 10 g 10g
1 options
Product Code. 30309298 Supplier Toronto Research Chemicals Supplier No. TRCA60249010G
 Restricted Item
Beräknad leverans: 07-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Amino-2-bromobenzoic Acid
CAS 2840-02-0
Molecular Formula C7H6NO2Br
Quantity 10 g
InChI Formula InChI=1S/C7H6BrNO2/c8-6-2-1-4(9)3-5(6)7(10)11/h1-3H,9H2,(H,10,11)
SMILES Nc1ccc(Br)c(c1)C(=O)O
IUPAC Name 5-amino-2-bromobenzoic acid
Molecular Weight (g/mol) 216.03
Formula Weight 214.9582