missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chloro-3-picoline, 97%, Thermo Scientific™

Catalog Number p-4380038
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
38186-82-2
C6H7ClN2
142.59
MFCD03095087
VSBISZPNLZFTPG-UHFFFAOYSA-N
5-amino-2-chloro-3-picoline, 2-chloro-3-methyl-5-aminopyridine, 5-amino-2-chloro-3-methylpyridine, 3-pyridinamine, 6-chloro-5-methyl, 6-chloro-5-methyl-3-pyridylamine, 6-chloro-5-methyl-pyridin-3-ylamine, 6-chloro-5-methylpyridine-3-amine, pubchem1152, 5-methyl-6-chloropyridine-3-amine, 6-chloro-5-methyl-3-pyridinamine
12295607
6-chloro-5-methylpyridin-3-amine
CC1=CC(N)=CN=C1Cl
This item is not returnable. View return policy
38186-82-2
C6H7ClN2
142.59
MFCD03095087
VSBISZPNLZFTPG-UHFFFAOYSA-N
5-amino-2-chloro-3-picoline, 2-chloro-3-methyl-5-aminopyridine, 5-amino-2-chloro-3-methylpyridine, 3-pyridinamine, 6-chloro-5-methyl, 6-chloro-5-methyl-3-pyridylamine, 6-chloro-5-methyl-pyridin-3-ylamine, 6-chloro-5-methylpyridine-3-amine, pubchem1152, 5-methyl-6-chloropyridine-3-amine, 6-chloro-5-methyl-3-pyridinamine
12295607
6-chloro-5-methylpyridin-3-amine
CC1=CC(N)=CN=C1Cl
Melting Point 90°C to 92°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 5-Amino-2-chloro-3-picoline
compliance-icons
Product Identifier
  • 5-Amino-2-chloro-3-picoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'