missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chloropyrimidine, 95%, Thermo Scientific Chemicals

Catalog Number p-4228307
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
56621-90-0
C4H4ClN3
129.55
DZBKIOJXVOECRA-UHFFFAOYSA-N
12215993
2-chloropyrimidin-5-amine
C1=C(C=NC(=N1)Cl)N
This item is not returnable. View return policy
56621-90-0
C4H4ClN3
129.55
DZBKIOJXVOECRA-UHFFFAOYSA-N
12215993
2-chloropyrimidin-5-amine
C1=C(C=NC(=N1)Cl)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 160°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Synonym 5-amino-2-chloropyrimidine, 2-chloro-pyrimidin-5-ylamine, 2-chloro-5-aminopyrimidine, 5-pyrimidinamine, 2-chloro, 2-chloropyrimidine-5-ylamine, pubchem6963, acmc-1axfh, 2-chloropyrimidin-5-ylamine, 5-pyrimidinamine,2-chloro, 5-amino-2-chloro pyrimidine
Formula Weight 129.55
Percent Purity 95%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 5-Amino-2-chloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-2-chloropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'