Läs mer
5-Amino-2-methylbenzenesulfonic Acid, TRC
Produktkod. 17913465
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17913465 2.5 g 2.5g
17920845 250 mg -
17940655 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17913465 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5773132.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 118-88-7
Molekylformel C7 H9 NO3 S
Molekylvikt (g/mol) 187.216
Synonym 1-Amino-4-methylbenzene-3-sulfonic Acid, 2-Methyl-5-amino benzene sulfonic Acid, 3-Amino-6-methylbenzenesulfonic Acid, 3-Sulfo-4-methylaniline, 4-Amino-2-sulfotoluene, 4-Aminotoluene-2-sulfonic Acid, 4-Methyl-3-sulfoaniline, 4-Methylaniline-3-sulfonic Acid, 4-Toluidine-3-sulfonic Acid, 5-Amino-2-methyl-1-sulfobenzene, 5-Amino-o-toluenesulfonic Acid, NSC 22958, NSC 5586, p-Toluidine-o-sulfonic Acid
IUPAC-namn 5-amino-2-methyl-benzenesulfonic acid
LEDER Cc1ccc(N)cc1S(=O)(=O)O
Specifikationer
Kemiskt namn eller material 5-Amino-2-methylbenzenesulfonic Acid
Kvantitet 2.5 g
InChI formel InChI=1S/C7H9NO3S/c1-5-2-3-6(8)4-7(5)12(9,10)11/h2-4H,8H2,1H3,(H,9,10,11)
Formel vikt 187.0303