Learn More
5-Amino-2-methylphenylboronic acid pinacol ester, 97%, Thermo Scientific™
Catalog Number p-4602014
Click to view available options
Quantity:
1 g
5 g
882670-69-1
C13H20BNO2
233.12
HPNBWHDRLCVZFV-UHFFFAOYSA-N
4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-amino-2-methylphenylboronic acid, pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, hpnbwhdrlcvzfv-uhfffaoysa-n, 5-amino-2-methyl-phenylboronic acid pinacol ester, 5-amino-2-methylphenyl boronic acid pinacol ester
16086089
882670-69-1
C13H20BNO2
233.12
HPNBWHDRLCVZFV-UHFFFAOYSA-N
4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-amino-2-methylphenylboronic acid, pinacol ester, 5-amino-2-methylphenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, hpnbwhdrlcvzfv-uhfffaoysa-n, 5-amino-2-methyl-phenylboronic acid pinacol ester, 5-amino-2-methylphenyl boronic acid pinacol ester
16086089
Melting Point 83°C to 84°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 233.12
Percent Purity 97%
Chemical Name or Material 5-Amino-2-methylphenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 5-Amino-2-methylphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.