Learn More
5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 3326-34-9 |
| Molecular Formula | C20H13NO5 |
| MDL Number | MFCD00005052 |
| Quantity | 250 mg |
| SMILES | O=C1OC2(C3=C(OC4=C2C=CC(O)=C4)C=C(O)C=C3)C5=C1C=C(N)C=C5 |
| Molecular Weight (g/mol) | 347.32 |
| Percent Purity | 96% |
| Chemical Name or Material | 5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one |