missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4H-1,2,4-triazole-3-thiol, tech., Thermo Scientific™

Catalog Number p-4602017
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
16691-43-3
C2H4N4S
116.14
MFCD00005231
WZUUZPAYWFIBDF-UHFFFAOYSA-N
3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
2723869
5-amino-1,2-dihydro-1,2,4-triazole-3-thione
NC1=NC(=S)NN1
This item is not returnable. View return policy
16691-43-3
C2H4N4S
116.14
MFCD00005231
WZUUZPAYWFIBDF-UHFFFAOYSA-N
3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
2723869
5-amino-1,2-dihydro-1,2,4-triazole-3-thione
NC1=NC(=S)NN1
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'