Läs mer
5-Amino-6-methylbenzimidazolone, TRC
Produktkod. 30135798
Change view
Kvantitet:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135798 1 g
30009082 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135798 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6135201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67014-36-2
Molekylformel C8 H9 N3 O
Molekylvikt (g/mol) 163.18
IUPAC-namn 5-amino-6-methyl-1,3-dihydrobenzimidazol-2-one
LEDER Cc1cc2NC(=O)Nc2cc1N
Specifikationer
Kemiskt namn eller material 5-Amino-6-methylbenzimidazolone
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C8H9N3O/c1-4-2-6-7(3-5(4)9)11-8(12)10-6/h2-3H,9H2,1H3,(H2,10,11,12)
Formel vikt 163.0746
Procent renhet >95%