Läs mer
5-Amino-N1-methyl-1H-Imidazole-1,4-dicarboxamide, Mikromol™
Produktkod. 30460405
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460405 4 x 25 mg
30049999 25 mg
30051751 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460405 Leverantör Mikromol Leverantörsnummer MM1480.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Amino-N1-methyl-1H-Imidazole-1,4-dicarboxamide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Amino-N1-methyl-1H-Imidazole-1,4-dicarboxamide
CAS 188612-53-5
Analyt- eller komponentnamn 5-Amino-N1-methyl-1H-Imidazole-1,4-dicarboxamide
Frakt skick Ice pack (-20°C)
Molekylformel C6 H9 N5 O2
Certifikat och efterlevnad ISO 17025
Synonym 5-Amino-N1-methyl-1H-Imidazole-1,4-dicarboxamide
InChI formel InChI=1S/C6H9N5O2/c1-9-6(13)11-2-10-3(4(11)7)5(8)12/h2H,7H2,1H3,(H2,8,12)(H,9,13)
Lösningstyp Neat
LEDER O=C(N)C=1N=CN(C(=O)NC)C1N
IUPAC-namn 5-amino-1-N-methylimidazole-1,4-dicarboxamide
Molekylvikt (g/mol) 183.17
Kvantitet 4 x 25 mg
Formel vikt 183.0756 g/mol