Läs mer
5-Amino-N1-methyl-1H-imidazole-1,4-dicarboxamide, TRC
Produktkod. 30132997
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132997 25 mg
30005628 5 mg
30010313 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132997 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61630025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 188612-53-5
Molekylformel C6 H9 N5 O2
Molekylvikt (g/mol) 183.17
Synonym 5-Amino-N1-methyl-1H-Imidazole-1,4-dicarboxamide
IUPAC-namn 5-amino-1-N-methylimidazole-1,4-dicarboxamide
LEDER CNC(=O)n1cnc(C(=O)N)c1N
Specifikationer
Kemiskt namn eller material 5-Amino-N1-methyl-1H-Imidazole-1,4-dicarboxamide
Kvantitet 25 mg
InChI formel InChI=1S/C6H9N5O2/c1-9-6(13)11-2-10-3(4(11)7)5(8)12/h2H,7H2,1H3,(H2,8,12)(H,9,13)
Formel vikt 183.0756