Läs mer
5-Aminolevulinic-13C5-15N Acid Hydrochloride, TRC
Produktkod. 30137058
Change view
Kvantitet:
0.5 mg
2.5 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137058 2.5 mg -
30132912 5 mg -
30010716 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137058 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6325882.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1196157-79-5
Molekylformel 13C5H915NO3 • HCl
Molekylvikt (g/mol) 137.093646
Synonym 5-Amino-4-oxopentanoic-13C5-15N Acid Hydrochloride, δ-Aminolevulinic-13C5-15N Acid Hydrochloride, Aminolevulinic-13C5-15N Acid Hydrochloride, 5-ALA-13C5-15N HCl, Levulan-13C5-15N
IUPAC-namn 5-(15N)azanyl-4-oxo(1,2,3,4,5-13C5)pentanoic acid;hydrochloride
LEDER [15NH2][13C]([H])([13C]([13C]([H])([13C]([H])([13C](O)=O)[H])[H])=O)[H].Cl
Specifikationer
Kemiskt namn eller material 5-Aminolevulinic-13C5-15N Acid Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C5H9NO3.ClH/c6-3-4(7)1-2-5(8)9;/h1-3,6H2,(H,8,9);1H/i1+1,2+1,3+1,4+1,5+1,6+1;
Procent renhet >95%