Learn More
5-(Aminomethyl)-2-methylpyrimidin-4-amine dihydrochloride, 95%, BLD Pharm™
Product Code. 31333215
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31333215 250 mg
31331257 5 g
31321200 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31333215 Supplier BLD Pharm Supplier No. BD106986250mg
 Restricted Item
Beräknad leverans: 25-11-2026
Specifications
CAS 874-43-1
Molecular Formula C6H12Cl2N4
MDL Number MFCD00058194
Quantity 250 mg
SMILES NC1=NC(C)=NC=C1CN.[H]Cl.[H]Cl
Molecular Weight (g/mol) 211.09
Percent Purity 95%
Chemical Name or Material 5-(Aminomethyl)-2-methylpyrimidin-4-amine dihydrochloride