Läs mer
5-Aminononane-5-carboxylic Acid, TRC
Produktkod. 30322185
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30322185 250 mg
1 options
Produktkod. 30322185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA615215250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Amino-2-butylhexanoic Acid
CAS 7597-66-2
Molekylformel C10 H21 N O2
Kvantitet 250 mg
Synonym Norleucine, 2-butyl- (9CI, ACI), 2-Butylnorleucine (ACI), 2-Amino-2-butylhexanoic acid, NSC 42318, α,α-Dibutylglycine, 5-Aminononane-5-carboxylic acid
InChI formel InChI=1S/C10H21NO2/c1-3-5-7-10(11,9(12)13)8-6-4-2/h3-8,11H2,1-2H3,(H,12,13)
LEDER CCCCC(N)(CCCC)C(=O)O
IUPAC-namn 2-amino-2-butylhexanoic acid
Molekylvikt (g/mol) 187.279
Formel vikt 187.157