Läs mer
5-Aminosalicylic Acid-13C6 Hydrochloride (Mesalazine-13C6 Hydrochloride), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Mesalazine-13C6 hydrochloride |
| CAS | 1261398-47-3 |
| Molekylformel | C 13C6 H7 N O3 . H Cl |
| Kvantitet | 10 mg |
| Synonym | 5-Amino-2-hydroxybenzoic-13C6 acid hydrochloride, 2-Hydroxy-5-aminobenzoic-13C6 acid hydrochloride, 5-Aminosalicylic acid-13C6 hydrochloride, 5-Aminosalicylic-1,2,3,4,5,6-13C6 acid hydrochloride, Mesalamine-13C6 hydrochloride, Mesalazine-13C6 hydrochloride, Mesalazine-13C6 (phenyl-13C6) hydrochloride, m-Aminosalicylic acid-13C6 hydrochloride |
| InChI formel | InChI=1S/C7H7NO3.ClH/c8-4-1-2-6(9)5(3-4)7(10)11;/h1-3,9H,8H2,(H,10,11);1H/i1+1,2+1,3+1,4+1,5+1,6+1; |
| LEDER | Cl.N[13c]1[13cH][13cH][13c](O)[13c]([13cH]1)C(=O)O |
| Molekylvikt (g/mol) | 195.55 |