Läs mer
5-Aminotetramethyl Rhodamine, TRC
Produktkod. 30318677
Change view
Kvantitet:
2 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30318677 2 mg
30315754 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30318677 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6303002MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Aminotetramethylrhodamine
CAS 167095-10-5
Molekylformel C24 H23 N3 O3
Kvantitet 2 mg
Synonym Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 5-amino-3',6'-bis(dimethylamino)-, 5-Aminotetramethyl Rhodamine
InChI formel InChI=1S/C24H23N3O3/c1-26(2)15-6-9-19-21(12-15)29-22-13-16(27(3)4)7-10-20(22)24(19)18-8-5-14(25)11-17(18)23(28)30-24/h5-13H,25H2,1-4H3
LEDER CN(C)c1ccc2c(Oc3cc(ccc3C24OC(=O)c5cc(N)ccc45)N(C)C)c1
IUPAC-namn 6-amino-3',6'-bis(dimethylamino)spiro[2-benzofuran-3,9'-xanthene]-1-one
Molekylvikt (g/mol) 401.46
Formel vikt 401.1739