Läs mer
5-Aza Cytosine-15N4, TRC
Produktkod. 30343724
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30343724 10 mg
30339100 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30343724 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79637710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Aza Cytosine-15N4
CAS 1189895-68-8
Molekylformel C3H415N4O
Kvantitet 10 mg
Synonym 5-Aza Cytosine-15N4, 4-Amino-s-triazin-2(1H)-one-15N4, 6-Amino-1,3,5-triazin-2(1H)-one-15N4, 4-Amino-s-triazin-2-ol-15N4, NSC 51100-15N4, NSC 54006-15N4
InChI formel InChI=1S/C3H4N4O/c4-2-5-1-6-3(8)7-2/h1H,(H3,4,5,6,7,8)/i4+1,5+1,6+1,7+1
LEDER [15NH2]C1=[15N]C([15NH]C=[15N]1)=O
IUPAC-namn 4-(amino-15N)-1,3,5-triazin-2(1H)-one-1,3,5-15N3
Molekylvikt (g/mol) 116.06
Formel vikt 116.03