Läs mer
5-Benzyloxy-2-mercaptobenzimidazole, TRC
Produktkod. 30333642
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30333642 100 mg
30333246 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30333642 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB285975100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Benzyloxy-2-mercaptobenzimidazole
CAS 465546-82-1
Molekylformel C14 H12 N2 O S
Kvantitet 100 mg
InChI formel InChI=1S/C14H12N2OS/c18-14-15-12-7-6-11(8-13(12)16-14)17-9-10-4-2-1-3-5-10/h1-8H,9H2,(H2,15,16,18)
LEDER Sc1nc2cc(OCc3ccccc3)ccc2[nH]1
IUPAC-namn 5-phenylmethoxy-1H-benzimidazole-2-thiol
Molekylvikt (g/mol) 256.32
Formel vikt 256.067