Läs mer
5-Beta-Pregnane-3-Alpha,20-Alpha-diol, TRC
Produktkod. 30006880
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006880 100 mg
17977164 10 mg
17989174 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006880 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP705130100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80-92-2
Molekylformel C21 H36 O2
Molekylvikt (g/mol) 320.51
Synonym Pregnanediol, 5beta-Pregnane-3alpha,20(S)-diol, 5beta-Pregnane-3alpha,20alph-diol, 3alpha,20alpha-Dihydroxy-5beta-pregnane, NSC 1612, NSC 47462
IUPAC-namn (3R,5R,8R,9S,10S,13S,14S,17S)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER C[C@H](O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Pregnanediol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C21H36O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h13-19,22-23H,4-12H2,1-3H3/t13-,14+,15+,16-,17+,18-,19-,20-,21+/m0/s1
Formel vikt 320.2715
Procent renhet >95%