Läs mer
5-[Bis(2-chloroethyl)amino]pyrimidine-2,4-diol, TRC
Produktkod. 30330638
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30330638 500 mg
1 options
Produktkod. 30330638 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB416270500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Uramustine
CAS 66-75-1
Molekylformel C8 H11 Cl2 N3 O2
Kvantitet 500 mg
Synonym 2,4(1H,3H)-Pyrimidinedione, 5-[bis(2-chloroethyl)amino]-, Uracil, 5-[bis(2-chloroethyl)amino]- (6CI,8CI), 5-[Bis(2-chloroethyl)amino]-2,4(1H,3H)-pyrimidinedione, 2,6-Dihydroxy-5-bis[2-chloroethyl]aminopyrimidine, 5-N,N-Bis(2-chloroethyl)aminouracil, 5-[Bis(2-chloroethyl)amino]uracil, 5-[Di(β-chloroethyl)amino]uracil, Aminouracil mustard, CB 4835, Chlorethaminacil, Demethyldopan, Desmethyldopan, ENT 50439, NCI C04820, NSC-34462, Nordopan, SK 19849, U-8344, Uracil mustard, Uracil nitrogen mustard, Uracillost, Uracilmostaza, Uramustin, Uramustine
InChI formel InChI=1S/C8H11Cl2N3O2/c9-1-3-13(4-2-10)6-5-11-8(15)12-7(6)14/h5H,1-4H2,(H2,11,12,14,15)
LEDER ClCCN(CCCl)C1=CNC(=O)NC1=O
IUPAC-namn 5-[bis(2-chloroethyl)amino]-1H-pyrimidine-2,4-dione
Molekylvikt (g/mol) 252.1
Formel vikt 251.0228