Learn More
Alfa Aesar™ 5-Bromo-1-indoleacetic acid, 95%
Brand:  Alfa Aesar™ H63039.03
Additional Details : CAS Number : 937621-97-1 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15479488
  • 2477.86 SEK / 1g
Estimated Shipment date
from Supplier 02-08-2018
Add to basket
Specifications
Chemical Identifiers
5-Bromo-1-indoleacetic acid
937621-97-1
MFCD09476492
22664359
Specifications
5-Bromo-1-indoleacetic acid
937621-97-1
Solid
C10H8BrNO2
22664359
C1=CC2=C(C=CN2CC(=O)O)C=C1Br
254.083
254.08
1g
95%
MFCD09476492
RVDSTBFKTGJQCS-UHFFFAOYSA-N
2-(5-bromoindol-1-yl)acetic acid
16495738