Learn More
5-Bromo-2,3-dichloropyridine, TRC
Product Code. 30337982
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30337982 1 g
1 options
Product Code. 30337982 Supplier Toronto Research Chemicals Supplier No. TRCB6911981G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 5-Bromo-2,3-dichloropyridine
CAS 97966-00-2
Molekylformel C5 H2 Br Cl2 N
Kvantitet 1 g
Synonym 2,3-Dichloro-5-bromopyridine, 3-Bromo-5,6-dichloropyridine
InChI formel InChI=1S/C5H2BrCl2N/c6-3-1-4(7)5(8)9-2-3/h1-2H
LEDER Clc1cc(Br)cnc1Cl
IUPAC-namn 5-bromo-2,3-dichloropyridine
Molekylvikt (g/mol) 226.886
Formel vikt 224.875