missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dimethoxypyrimidine, 97%, Thermo Scientific™

Catalog Number p-4323376
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
56686-16-9
QEZIMQMMEGPYTR-UHFFFAOYSA-N
2,4-dimethoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxy-1,3-diazine, 5-bromo-2,4-bis methyloxy pyrimidine, 5-bromo-2,4-dimethoxy-pyrimidine, pubchem5281, acmc-209lu1, ksc495s0l, 5-bromo-2,4-dimethoxypyrimidine, 5-bromo-2,4-dimethoxypyrimidine;
255719
5-bromo-2,4-dimethoxypyrimidine
COC1=NC(=NC=C1Br)OC
This item is not returnable. View return policy
56686-16-9
QEZIMQMMEGPYTR-UHFFFAOYSA-N
2,4-dimethoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxy-1,3-diazine, 5-bromo-2,4-bis methyloxy pyrimidine, 5-bromo-2,4-dimethoxy-pyrimidine, pubchem5281, acmc-209lu1, ksc495s0l, 5-bromo-2,4-dimethoxypyrimidine, 5-bromo-2,4-dimethoxypyrimidine;
255719
5-bromo-2,4-dimethoxypyrimidine
COC1=NC(=NC=C1Br)OC
CAS Min % 96.0
CAS Max % 100.0
Melting Point 62°C to 67°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H7BrN2O2
Quantity 1 g
Molecular Weight (g/mol) 219.038
Formula Weight 219.04
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 5-Bromo-2, 4-dimethoxypyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2,4-dimethoxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'