missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-6-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055690
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
132606-40-7
C6H5BrClN
206.47
MFCD07375117
DTFBHJWQTDQBEM-UHFFFAOYSA-N
5-bromo-2-chloro-6-methylpyridine, 3-bromo-6-chloro-2-picoline, 3-bromo-6-chloro-2-methyl-pyridine, 5-bromo-2-chloro-6-picoline, 2-chloro-5-bromo-6-methylpyridine, 2-chloro-6-methyl-5-bromopyridine, pyridine, 3-bromo-6-chloro-2-methyl, 2-chloro-5-bromo-6-picoline, pubchem6208
14866674
3-bromo-6-chloro-2-methylpyridine
CC1=C(Br)C=CC(Cl)=N1
This item is not returnable. View return policy
132606-40-7
C6H5BrClN
206.47
MFCD07375117
DTFBHJWQTDQBEM-UHFFFAOYSA-N
5-bromo-2-chloro-6-methylpyridine, 3-bromo-6-chloro-2-picoline, 3-bromo-6-chloro-2-methyl-pyridine, 5-bromo-2-chloro-6-picoline, 2-chloro-5-bromo-6-methylpyridine, 2-chloro-6-methyl-5-bromopyridine, pyridine, 3-bromo-6-chloro-2-methyl, 2-chloro-5-bromo-6-picoline, pubchem6208
14866674
3-bromo-6-chloro-2-methylpyridine
CC1=C(Br)C=CC(Cl)=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 46°C to 47°C
Boiling Point 80°C to 84°C (2 mmHg)(literature)
Quantity 5 g
Beilstein 4243997
Formula Weight 206.47
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-chloro-6-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'