missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-cyanobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H61273.03

Additional Details : CAS Number : 1032231-30-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497778

  • 1071.80 SEK / 1g


Estimated Shipment date
from Supplier 21-02-2019
Add to basket

Specifications

Chemical Identifiers

1032231-30-3
225.836
5-bromo-2-cyanophenyl boronic acid, 2-cyano-5-bromophenylboronic acid, 5-bromo-2-cyanophenyl boronicacid, 5-bromo-2-cyano-phenyl boronic acid
(5-bromo-2-cyanophenyl)boronic acid
C7H5BBrNO2
BVCIFFSGTUEHOP-UHFFFAOYSA-N
46738962
B(C1=C(C=CC(=C1)Br)C#N)(O)O

Specifications

5-Bromo-2-cyanobenzeneboronic acid
1032231-30-3
5-bromo-2-cyanophenyl boronic acid, 2-cyano-5-bromophenylboronic acid, 5-bromo-2-cyanophenyl boronicacid, 5-bromo-2-cyano-phenyl boronic acid
B(C1=C(C=CC(=C1)Br)C#N)(O)O
225.836
225.84
97%
1g
C7H5BBrNO2
BVCIFFSGTUEHOP-UHFFFAOYSA-N
(5-bromo-2-cyanophenyl)boronic acid
46738962
Solid
Videos
SDS