missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-cyanopyrimidine, 95%, Thermo Scientific Chemicals

Catalog Number p-7053729
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
38275-57-9
C5H2BrN3
183.996
MFCD02940446
VPQICCOHFSGBMA-UHFFFAOYSA-N
5-bromo-2-cyanopyrimidine, 5-bromo-2-pyrimidinecarbonitrile, 2-pyrimidinecarbonitrile, 5-bromo, 5-bromo-pyrimidine-2-carbonitile, 2-cyano-5-bromopyrimidine, pubchem5278, acmc-1ad2b, 5-bromo-2-cyano-pyrimidine, ksc497m1f, 5-bromo-pyrimidine-2-carbonitrile
686546
5-bromopyrimidine-2-carbonitrile
C1=C(C=NC(=N1)C#N)Br
This item is not returnable. View return policy
38275-57-9
C5H2BrN3
183.996
MFCD02940446
VPQICCOHFSGBMA-UHFFFAOYSA-N
5-bromo-2-cyanopyrimidine, 5-bromo-2-pyrimidinecarbonitrile, 2-pyrimidinecarbonitrile, 5-bromo, 5-bromo-pyrimidine-2-carbonitile, 2-cyano-5-bromopyrimidine, pubchem5278, acmc-1ad2b, 5-bromo-2-cyano-pyrimidine, ksc497m1f, 5-bromo-pyrimidine-2-carbonitrile
686546
5-bromopyrimidine-2-carbonitrile
C1=C(C=NC(=N1)C#N)Br

5-Bromo-2-cyanopyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-cyanopyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture.
TRUSTED_SUSTAINABILITY
Color White to Yellow
Quantity 1 g
UN Number UN3439
Beilstein 775324
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 183.99
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-cyanopyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'