missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-ethoxypyridine, 97%, Thermo Scientific™

Catalog Number p-4258805
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
55849-30-4
C7H8BrNO
202.05
WQXZKMUZWPUZGL-UHFFFAOYSA-N
2-ethoxy-5-bromopyridine, 3-bromo-6-ethoxypyridine, 5-bromo-2-ethoxy-pyridine, pyridine, 5-bromo-2-ethoxy, zlchem 405, pubchem6235, 5-bromo-ethoxy-pyridine, acmc-1avox, 5-bromo-2-ethoxy pyridine, ksc497o6h
817237
5-bromo-2-ethoxypyridine
CCOC1=NC=C(C=C1)Br
This item is not returnable. View return policy
55849-30-4
C7H8BrNO
202.05
WQXZKMUZWPUZGL-UHFFFAOYSA-N
2-ethoxy-5-bromopyridine, 3-bromo-6-ethoxypyridine, 5-bromo-2-ethoxy-pyridine, pyridine, 5-bromo-2-ethoxy, zlchem 405, pubchem6235, 5-bromo-ethoxy-pyridine, acmc-1avox, 5-bromo-2-ethoxy pyridine, ksc497o6h
817237
5-bromo-2-ethoxypyridine
CCOC1=NC=C(C=C1)Br
CAS Min % 96.0
CAS Max % 100.0
Melting Point 34°C to 37°C
Color White to Yellow
Boiling Point 107°C (33.0 Torr)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 202.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-ethoxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-ethoxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'