missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzamide, 96%, Thermo Scientific™

Catalog Number p-7054335
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
214210-17-0
C7H5BrFNO
218.03
MFCD03094430
MAGQVUXQRXLNGB-UHFFFAOYSA-N
5-bromo-2-fluoro-benzamide, 2-fluoro-5-bromobenzamide
2778901
5-bromo-2-fluorobenzamide
NC(=O)C1=C(F)C=CC(Br)=C1
This item is not returnable. View return policy
214210-17-0
C7H5BrFNO
218.03
MFCD03094430
MAGQVUXQRXLNGB-UHFFFAOYSA-N
5-bromo-2-fluoro-benzamide, 2-fluoro-5-bromobenzamide
2778901
5-bromo-2-fluorobenzamide
NC(=O)C1=C(F)C=CC(Br)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 218.02
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-fluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'