Learn More
5-Bromo-2-hydroxy-4-methylpyridine, 97%, Thermo Scientific Chemicals
Catalog Number p-7055954
Click to view available options
Quantity:
5 g
25 g
164513-38-6
C6H6BrNO
188.024
MFCD03427664
BYGNXKDNIFHZHQ-UHFFFAOYSA-N
5-bromo-2-hydroxy-4-methylpyridine, 5-bromo-4-methylpyridin-2-ol, 5-bromo-2-hydroxy-4-picoline, 5-bromo-4-methyl-2 1h-pyridinone, 5-bromo-4-methyl-2-pyridinol, 2-hydroxy-4-methyl-5-bromopyridine, 5-bromo-4-methyl-pyridin-2-ol, pubchem6564, acmc-209dre, ksc496a8d
819937
5-bromo-4-methyl-1H-pyridin-2-one
CC1=CC(=O)NC=C1Br
164513-38-6
C6H6BrNO
188.024
MFCD03427664
BYGNXKDNIFHZHQ-UHFFFAOYSA-N
5-bromo-2-hydroxy-4-methylpyridine, 5-bromo-4-methylpyridin-2-ol, 5-bromo-2-hydroxy-4-picoline, 5-bromo-4-methyl-2 1h-pyridinone, 5-bromo-4-methyl-2-pyridinol, 2-hydroxy-4-methyl-5-bromopyridine, 5-bromo-4-methyl-pyridin-2-ol, pubchem6564, acmc-209dre, ksc496a8d
819937
5-bromo-4-methyl-1H-pyridin-2-one
CC1=CC(=O)NC=C1Br
It is used for medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used for medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 198°C to 202°C
Color White
Quantity 5 g
Beilstein 6798531
Solubility Information Slightly soluble in water.
Formula Weight 188.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-hydroxy-4-methylpyridine