Läs mer
5-Bromo-2-hydroxypyrimidine, 98%, Thermo Scientific™
Produktkod. 11304965
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11304965 1 g 1g
11314965 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11304965 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L18347.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38353-06-9
Molekylformel C4H3BrN2O
Molekylvikt (g/mol) 174.985
MDL-nummer MFCD00233952
InChI-nyckel VTUDATOSQGYWML-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxypyrimidine, 5-bromopyrimidin-2-ol, 5-bromo-2-pyrimidinol, 5-bromo-2-pyrimidinone, 2 1h-pyrimidinone, 5-bromo, 5-bromopyrimidin-2 1h-one, 5-bromo-2 1h-pyrimidinone, 5-bromopyrimidin-2-one, 5-bromo-pyrimidin-2-ol, 2-pyrimidinol, 5-bromo
PubChem CID 101494
IUPAC-namn 5-bromo-1H-pyrimidin-2-one
LEDER C1=C(C=NC(=O)N1)Br
Specifikationer
Kemiskt namn eller material 5-Bromo-2-hydroxypyrimidine
Smältpunkt ∽230°C (decomposition)
Kvantitet 1 g
Beilstein 742202
Formel vikt 174.99
Procent renhet 98%