Learn More
5-Bromo-2-isopropoxybenzonitrile, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Quantity | 250 mg |
UN Number | UN3439 |
Formula Weight | 240.1 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-2-isopropoxybenzonitrile |