missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylindole, 98%, Thermo Scientific Chemicals

Catalog Number p-4258811
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1075-34-9
C9H8BrN
210.07
BJUZAZKEDCDGRW-UHFFFAOYSA-N
5003968
5-bromo-2-methyl-1H-indole
CC1=CC2=C(N1)C=CC(=C2)Br
This item is not returnable. View return policy
1075-34-9
C9H8BrN
210.07
BJUZAZKEDCDGRW-UHFFFAOYSA-N
5003968
5-bromo-2-methyl-1H-indole
CC1=CC2=C(N1)C=CC(=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 104.0°C to 106.0°C
Color Tan to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 5-bromo-2-methylindole, 2-methyl-5-bromoindole, 1h-indole, 5-bromo-2-methyl, 5-bromo-2-methylindol, pubchem9382, acmc-209xly, 5-bromo-2-methyl-indole, 2-methyl-5-bromo-1h-indole, 1h-indole,5-bromo-2-methyl
Formula Weight 210.07
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromo-2-methylindole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methylindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'