missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-(trifluoromethoxy)benzaldehyde, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
Description
5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
Specifications
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 269.01 |
Percent Purity | 98% |
Chemical Name or Material | 5-Bromo-2-(trifluoromethoxy)benzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'