Learn More
5-Bromo-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Catalog Number p-4813194
Click to view available options
Quantity:
500 mg
5 g
436799-32-5
C6H3BrF3N
226
RPFAUCIXZGMCFN-UHFFFAOYSA-N
5-bromo-2-trifluoromethyl pyridine, 2-trifluoromethyl-5-bromopyridine, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromopyridine, trifluoromethyl-5-bromo-2-pyridine, 5-bromo-2-trifluoromethyl-pyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, pubchem2164
2761197
5-bromo-2-(trifluoromethyl)pyridine
C1=CC(=NC=C1Br)C(F)(F)F
436799-32-5
C6H3BrF3N
226
RPFAUCIXZGMCFN-UHFFFAOYSA-N
5-bromo-2-trifluoromethyl pyridine, 2-trifluoromethyl-5-bromopyridine, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromopyridine, trifluoromethyl-5-bromo-2-pyridine, 5-bromo-2-trifluoromethyl-pyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, pubchem2164
2761197
5-bromo-2-(trifluoromethyl)pyridine
C1=CC(=NC=C1Br)C(F)(F)F
Melting Point 42.0°C to 43.0°C
Flash Point 78°C
Assay Percent Range 96% min. (GC)
Quantity 500 mg
Formula Weight 226
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine
compliance-icons
Product Identifier
  • 5-Bromo-2-(trifluoromethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.