Learn More
Alfa Aesar™ 5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridyl)indole, 97%
Brand:  Alfa Aesar™ H63282.03
Additional Details : CAS Number : 127792-80-7 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15448216
  • 2256.11 SEK / 1g
Estimated Shipment date
from Supplier 12-03-2018
Add to basket
Specifications
Chemical Identifiers
5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
277.16
4184918
127792-80-7
MFCD07366543
Specifications
5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
190° to 192°C
Solid
277.16
4184918
127792-80-7
1g
97%
MFCD07366543