missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole, 97%, Alfa Aesar™

Brand:  Alfa Aesar H63590.03

Additional Details : CAS Number : 282546-99-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498386

  • 1634.23 SEK / 1g


Estimated Shipment date
from Supplier 17-04-2019
Add to basket

Specifications

Chemical Identifiers

282546-99-0
334.213
ZGKVTJQGVWHCDH-UHFFFAOYSA-N
2763989
C1CC2(CC=C1C3=CNC4=C3C=C(C=C4)Br)OCCO2
C16H16BrNO2
MFCD06411613
5-bromo-3-1,4-dioxaspiro 4.5 dec-7-en-8-yl-1h-indole, 5-bromo-3-1,4-dioxaspiro 4.5 dec-7-en-8-yl-1h-indole, 5-bromo-3-1,4-dioxaspiro 4,5 dec-7-en-8-yl-1h-indole, bromodioxaspirodecenylindole, 3-1,4-dioxa-spiro 4,5 dec-7-en-8-yl-5-bromo-1h-indole, 5-bromo-3-1,4-dioxaspiro 4.5 dec-8-en-8-yl-1h-indole
5-bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole

Specifications

5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
282546-99-0
MFCD06411613
5-bromo-3-1,4-dioxaspiro 4.5 dec-7-en-8-yl-1h-indole, 5-bromo-3-1,4-dioxaspiro 4.5 dec-7-en-8-yl-1h-indole, 5-bromo-3-1,4-dioxaspiro 4,5 dec-7-en-8-yl-1h-indole, bromodioxaspirodecenylindole, 3-1,4-dioxa-spiro 4,5 dec-7-en-8-yl-5-bromo-1h-indole, 5-bromo-3-1,4-dioxaspiro 4.5 dec-8-en-8-yl-1h-indole
C1CC2(CC=C1C3=CNC4=C3C=C(C=C4)Br)OCCO2
334.213
334.21
97%
1g
C16H16BrNO2
14255489
ZGKVTJQGVWHCDH-UHFFFAOYSA-N
5-bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole
2763989
Solid
Videos
SDS