missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Bromo-3-(1-piperidinylmethyl)indole, 95%
Brand: Alfa Aesar™ H63087.06
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
5-Bromo-3-(1-piperidinylmethyl)indole | |
293.2 | |
MFCD28126772 |
Specifications
5-Bromo-3-(1-piperidinylmethyl)indole | |
Solid | |
293.2 |
5g | |
95% | |
MFCD28126772 |