Läs mer
5-Bromo-3-[[(2S)-1-methyl-2-pyrrolidinyl]methyl]-1H-indole, TRC
Produktkod. 30346485
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30346485 10 mg
30338548 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30346485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68872510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Bromo-3-(((2S)-1-methyl-2-pyrrolidinyl)methyl)-1H-indole
CAS 208464-41-9
Molekylformel C23 H23 N3 O13 S
Kvantitet 10 mg
Synonym 5-Bromo-3-[[(2S)-1-Methyl-2-Pyrrolidinyl]Methyl]-1H-Indole Impurity
InChI formel InChI=1S/C23H23N3O13S/c1-10(27)35-16-17(36-11(2)28)19(37-12(3)29)22(39-18(16)21(31)34-4)38-14-8-6-5-7-13(14)20(30)25-23-24-9-15(40-23)26(32)33/h5-9,16-19,22H,1-4H3,(H,24,25,30)/t16-,17?,18?,19?,22-/m0/s1
LEDER COC(=O)C1O[C@H](Oc2ccccc2C(=O)Nc3ncc(s3)[N+](=O)[O-])C(OC(=O)C)C(OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn 5-bromo-3-[[(2S)-1-methylpyrrolidin-2-yl]methyl]-1H-indole
Molekylvikt (g/mol) 581.506
Formel vikt 581.095